Skip to Content
Merck
All Photos(1)

Key Documents

1.46624

Millipore

FRIIS Liquid Medium

pack of 1 × 800 mL, nonselective for mycoplasma

Synonym(s):

FRIIS Broth, Mycoplasma Broth, Mycoplasma Liquid Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
Pricing and availability is not currently available.

Agency

EP 2.6.7

Quality Level

form

liquid

feature

ready-to-use

packaging

pack of 1 × 800 mL

technique(s)

microbiological culture: suitable

pH range

7.6-7.8

application(s)

pharmaceutical

storage temp.

2-8°C

suitability

nonselective for mycoplasma


General description

FRIIS liquid Medium is designed for qualitative detection of avian mycoplasmas in test samples of pharmaceutical industry especially in bulk vaccines, cell banks and viral cultures.
The media formulation is basic and is supplemented NAD (Nucleotide Adenine Dinucleotide) which supports the growth M. synoviae. The color change reaction in the medium indicates the presence of mycoplasmas in the tested sample. Addition of antibiotics in the medium inhibits the accompanying microflora.

Application

FRIIS liquid Medium is designed for the detection of avian mycoplasmas in test samples of pharmaceutical industry especially in bulk vaccines, cell banks and viral cultures.

Analysis Note

The appearance of the medium is clear and red and color change reaction from red to yellow determines the presence of mycoplasma.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service