Skip to Content
Merck
All Photos(1)

Key Documents

1.46567

Millipore

Tryptic Soy Broth Ready-to-use Bottles

Tween® 1 %, bottle capacity 1000 mL, bottle filling volume 850 mL, closure type, red screw cap, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth with 1% Polysorbate concentrated, SCDB, Soyabean Casein Digest Broth, TSB, TSB with Neutralizer, Tryptic Soy Broth (TSB), Tryptic Soy Broth with 1% Tween concentrated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.13
JP 4.06
USP 71

Quality Level

feature

closure type red screw cap
ready-to-use

composition

Tween®, 1%

packaging

box of 6 bottles

bottle capacity

1000 mL

bottle filling volume

850 mL

capacity

1000 mL

pH

7.3

application(s)

food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth (TSB) with Tween®80 is a complex medium designed for the enumeration of total microbial count present in the test sample. Polysorbate 80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, di-chlorobenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrol (PHB esters).
Upon incubation, the enumerated microflora can be plated, isolated, and identified.

Application

Tryptic Soy Broth (TSB) Tween®80 is a complex medium used to enumerate microorganisms present in sterile or non-sterile products from food or pharmaceutical industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service