Skip to Content
Merck
All Photos(1)

Documents

1.46191

Millipore

Malt Extract Agar Contact

plate size 55 mm, for yeasts, for molds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

ISO

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

ready-to-use

plate size

55 mm

pH

5.6 ( in H2O)

application(s)

cosmetics
food and beverages
pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

15-25°C

suitability

molds
yeasts

General description

Malt Extract Agar Contact - RT is designed for environmental monitoring in the non-critical area for the detection of yeasts and molds.
Malt Extract Agar is used for the detection of yeasts and molds. The pH of 5.4 - 5.8 of the medium promotes the growth of most yeasts and fungi, while bacterial growth is more or less inhibited. Malt extract has a high content of carbohydrates (90-92 %), nitrogen compounds, and vitamins, e.g., riboflavin, pyridoxin, and thiamine. The soymeal peptone is also rich in carbohydrates, and additionally supplies the necessary protein compounds such as peptides and amino acids.

Application

The Malt Extract Agar is used for the identification of yeasts and molds in food, including milk and dairy products, and environmental samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service