Skip to Content
Merck
All Photos(1)

Documents

1.46153

Millipore

TSA Cont. + Penase+LTHTh-ICR

Sign Into View Organizational & Contract Pricing


About This Item

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated by SER-TAIN process

form

solid

feature

2D bar code
ready-to-use

D

55 mm

pH

7.3 ( in H2O)

application(s)

microbiology
pharmaceutical
surface monitoring

storage temp.

20-25°C

suitability

bacteria
molds
yeasts

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service