Skip to Content
Merck
All Photos(1)

Key Documents

14-591-M

Sigma-Aldrich

SHP-1 Protein, 10 µg

N-terminal GST-tagged recombinant human full-length SHP-1, for use in Phosphatase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00

€578.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 μG
€578.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€578.00


Estimated to ship on30 May 2025


biological source

human

Quality Level

100
300

recombinant

expressed in E. coli

mol wt

Mw 95 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST-tagged recombinant human full-length SHP-1
Product Source: expressed in E. coli

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: SHP-1
Target Sub-Family: Tyr phosphatase

Quality

routinely evaluated by dephosphorylation of 100mM DiFMUP

Physical form

Glutathione-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I'd like to know how much concentration the following product is. 14-591-M Sigma-Aldrich SHP-1 Protein, 10 µg

    1 answer
    1. The concentration of this product is lot-specific. Historically, lots tend to have a concentration of approximately 0.5 mg/mL.

      Kindly review the previous certificate of analyses under the Supporting Documentation tab available at this link: https://www.emdmillipore.com/US/en/product/SHP-1-Protein-10-g,MM_NF-14-591#documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service