Skip to Content
Merck
All Photos(1)

Key Documents

14-504-M

Sigma-Aldrich

PKCδ Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant amino acids 2-end, of human PKCδ. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00

€578.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€578.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€578.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

human

Quality Level

100
300

recombinant

expressed in Sf21 cells

mol wt

Mw 78.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PRKCD(5580)

General description

N-terminal His6-tagged, recombinant amino acids 2-end, of human PKCδ

PKC Lipid activator (Catalog # 20-133) is required for use.
Product Source: Human PKCδ expressed in Sf21 cells.

Biochem/physiol Actions

Protein Target: PKCδ

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-504M when ordering the 250 µg size.

Quality

Routinely evaluated by phosphorylation of PKC Substrate Peptide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service