Skip to Content
Merck

1.37117

SAFC

Dimethyl sulfoxide

EMPROVE® EXPERT, Ph. Eur., ChP, USP

Pharma Manufacturing

Synonym(s):

Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
12352119
EC Index Number:
200-664-3

Agency

ChP
Ph. Eur.
USP

Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

product line

EMPROVE® EXPERT

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.479 (lit.)

kinematic viscosity

2.14 cSt

bp

189 °C (lit.)

mp

16-19 °C (lit.)

transition temp

flash point 87 °C

density

1.10 g/mL (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

storage temp.

2-30°C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Dimethyl sulfoxide is used as penetration enhancer in topical formulation, as cryopreservative in cell therapy and solvent in antibody drug conjugate production.
Due to its low microbial and endotoxin limits, Dimethyl Sulfoxide Emprove® Expert is especially suitable for high-risk applications.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Specifications

8.0 g/m3 (20 °C)

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understand how considerations such as DMSO concentrations and freezing techniques are key in the successful implementation of high cell density cryopreservation (HCDC).

Find best practices for using Pellicon® Capsules in ADC processing as well as data from two studies using Pellicon® Capsules and Pellicon® cassettes in the ADC manufacturing process.

Related Content

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service