Skip to Content
Merck
All Photos(1)

Key Documents

1.25167

Millipore

Eshmuno® CPS Resin

RoboColumn Eshmuno® CPS, volume 0.6 mL, prepacked column

Synonym(s):

RoboColumn Eshmuno® CPS, 0.6ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€343.00

€343.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 EA
€343.00

About This Item

UNSPSC Code:
41115711

€343.00


Estimated to ship on29 May 2025


ligand

(Sulfoisobutyl)

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

packaging

pkg of 8 pieces

parameter

≤8 bar pressure
500 cm/hr max. flow rate

column I.D. × L

5 mm × 30 mm

volume

0.6 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~160 mL, resin binding capacity

application(s)

recombinant protein

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

8PC 5x30mm

Features and Benefits

Eshmuno® CPS chromatography resin enables:

  • Outstanding binding capacities at elevated conductivity levels (≥ 10 mS/cm)
  • Excellent separation efficiencies at high protein loadings
  • Ease of implementation and selection of process parameters through strong cation exchanger chemistry without hydrophobic groups
  • Established Fab fragments, recombinant human growth factor, and recombinant proteins purification protocols
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities
  • Robust and safe packing procedures
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service