Skip to Content
Merck
All Photos(1)

Documents

1.25140

Millipore

RoboColumn Eshmuno® S, 0.6 ml

column I.D. × L 5 mm × 30 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

(Sulfoisobutyl)

Quality Level

material

polypropylene housing

product line

Eshmuno®

parameter

≤8 bar pressure
1000 cm/hr max. flow rate

column I.D. × L

5 mm × 30 mm

volume

0.6 mL

matrix active group

Polyvinyl Ether

mean particle size

85 μm

capacity

~165 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

15-25°C

General description

5 x 30 mm

Features and Benefits

Eshmuno® S chromatography resin enables:

  • Purification of basic and neutral proteins (e.g., antibodies) and peptides specifically designed for highly productive downstream bioprocessing
  • High flow rate applications
  • High separation efficiency over a broad range of flow rates with a rigid matrix and optimized pore structure
  • Tentacle technology which provides high yield, selectivity, and focused elution
  • Multiple CIP and packing cycles with the chemically and physically robust bead, leading to a longer resin lifetime
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service