Skip to Content
Merck
All Photos(1)

Key Documents

1.20594

Millipore

PharmPrep® P100 RP-8e, 10 μm Scout Column

L × diam. 250 mm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
Pricing and availability is not currently available.

material

spherical silica gel

product line

PharmPrep®

parameter

≤40 bar max. pressure

L × diam.

250 mm × 4.6 mm

matrix active group

Spherical Silica Gel

pore size

10 nm mean pore size

application(s)

plasma purification

separation technique

reversed phase

General description

Spherical porous Silica with uniform and homogeneous gel matrix with excellent batch to batch reproducibility

Features and Benefits

PharmPrep® P100 RP-8e, 10 µm Scout Column provides users with:
  • Uniform and homogenous silica gel matrix with excellent batch-to-batch reproducibility over many column packings
  • Narrow particle size distribution for high performance and high productivity in small peptide purification processes
  • Reproducible specific surface area and pore size distribution
  • Low back pressure
  • Enhanced mechanical stability for long sorbent life-time
  • perfect separation of insulin and the main impurities desamido-insulin.

Legal Information

PHARMPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service