Skip to Content
Merck
All Photos(1)

Documents

1.19301

Supelco

Reference Standard

traceable to NIST SRM 2032, 935a (for chlorine, chlorine dioxide, ozone), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Agency

traceable to NIST SRM 2032, 935a (for chlorine, chlorine dioxide, ozone)

Quality Level

product line

Spectroquant®

specific analyte(s)

ozone

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move DC

storage temp.

15-25°C

General description

for chlorine, chlorine dioxide, ozone Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service