Skip to Content
Merck
All Photos(1)

Key Documents

1.19161

Supelco

Standard samples for HPLC system qualification

precision, methyl paraben in methanol 60 mg/L, for use with LICHROTEST

Synonym(s):

TM=["LICHROTEST"] Standard samples for HPLC system qualification Set 7: Precision

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€273.00

€273.00


Estimated to ship on25 March 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
€273.00

About This Item

UNSPSC Code:
41115709
NACRES:
NA.24

€273.00


Estimated to ship on25 March 2025


Request a Bulk Order

grade

analytical standard

Quality Level

composition

methyl paraben in methanol, 60 mg/L

technique(s)

HPLC: suitable

compatibility

for use with LICHROTEST

format

mixture

storage temp.

15-25°C

General description

Set 7: Precision; 60 mg/l Methyl paraben in methanol LiChroTest

Features and Benefits

  • Gravimetrically and volumetrically prepared standards using ISO 9001 quality process
  • Photometrically qualified standards concerning content using a high-performance spectrophotometer calibrated using NIST Standards

Analysis Note

60 mg/l Methyl paraben: in accordance

Legal Information

LICHROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service