Skip to Content
Merck
All Photos(1)

Key Documents

1.17243

Supelco

Fluoride Test, photometric

photometric, 0.10-2.50 mg/L (F-), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€116.00

€116.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
€116.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€116.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Fluoride Cell Test, photometric, 0.10-2.50 mg/L (F-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

fluoride

measuring range

0.10-2.50 mg/L (F-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (SPADNS method (As free))

storage temp.

15-25°C

General description

Method: photometric, SPADNS (Arsenic free) 0.10 - 2.50 mg/l Fˉ Spectroquants®

Fluoride ions react with part of an arsenic-free red zirconium-dye solution to form a colorless complex, which in turn results in a bleaching of the red color (SPADNS method). The concentration of the unconsumed zirconium-dye solution is subsequently determined photometrically.
The method is analogous to APHA 4500-F- D.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) , which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service