Skip to Content
Merck
All Photos(1)

Key Documents

1.16973

Supelco

Nitrite Test Strips, reflectometric

0.5-25.0 mg/L (NO₂⁻), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Nitrite chemical test strips, Nitrite test strip for instrument reader

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
€48.10

€48.10


Estimated to ship on31 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
€48.10

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€48.10


Estimated to ship on31 March 2025


Request a Bulk OrderRequest more information

Product Name

Nitrite Test, reflectometric, 0.5-25.0 mg/L (NO2-), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

measuring range

0.5-25.0 mg/L (NO2-)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 0.5 - 25.0 mg/l NO₂⁻ Reflectoquant®
Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems. On the Reflectoquant Nitrite test strips, in the presence of an acidic buffer nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts to form a red-violet dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the nitrite concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for nitrite. Further quantitative determination methods can be found with our Spectroquant tests.

Linkage

Replaces: 16973-1; 16973

Analysis Note

function control: conforms
Assay
- standard 1:
1.8 - 2.2 mg/l
- standard 2: 9.0 - 11.0 mg/l
- standard 3: 18.0 - 22.0 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service