Skip to Content
Merck

1.16886

Millipore

Fractogel® EMD COO- (M)

Synonym(s):

Fractogel® EMD COO⁻ (M)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

carboxymethyl

Quality Level

description

weak cation exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm)

sterility

sterile (Caustic Stable)

product line

Fractogel®

form

resin

specific analyte(s)

MAbs

parameter

300 cm/hr flow rate
8 bar max. pressure

matrix active group

methacrylate

mean particle size

40-90 μm

capacity

100 mg binding capacity (lysozyme/ml of resin)

transition temp

flash point 35 °C (calculated)

density

1.43 g/cm3 at 20 °C

application(s)

recombinant protein
vaccine development

separation technique

cation exchange
weak cation exchange

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Weak cation exchange Fractogel® resin featuring the tentacle technology, efficient for difficult aggregate removal

Features and Benefits

Fractogel® EMD COO- (M) enables:
  • Good resolution between monomers and aggregates
  • Efficient aggregate and HCP removal combined with high yield
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution


Due to the titration behavior, the ion exchange capacity can be used from pH 6 to pH 12. The separation of proteins is based on reversible electrostatic interactions between the negatively charged regions of the proteins′ surface and the support. Proteins are retained efficiently on Fractogel® EMD COO- (M) when the pH of the buffer is about 1 unit below their isoelectric points (pl).

The strength of the binding depends on the following:
  • the buffer system
  • pH value of the buffer which determines the surface charge of the protein
  • the degree of the ionization of the functional groups of the exchanger
  • the concentration of the counter ions
  • the charge density on the support (protein binding capacity)

Packaging

  • 1.16886.0100: Fractogel® EMD COO- (M) Resin 100ml
  • 1.16886.0010: Fractogel® EMD COO- (M) Resin 10ml
  • 1.16886.0500: Fractogel® EMD COO- (M) Resin 500ml
  • 1.16886.5000: Fractogel® EMD COO- (M) Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension,free from impurities(foreign particles)
Microscopic evaluation: Uniform spherical particles,no agglomerates,no fines
Extractable matter (water): ≤ 0.05 %
Cerium: ≤ 10 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Particle size (d10): 37 - 45 µm
Particle size (d50): 48 - 60 µm
Particle size (d90): 63 - 77 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Protein binding capacity (lysozyme): 80 - 120 mg/ml
Functional test (b:a): ≤ 0.30
Functional test (c:d): ≤ 0.70
Functional test: Separation of chymotrypsinogen A, cytochrome C and lysozyme

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service