Skip to Content
Merck
All Photos(4)

Key Documents

1.16738

Supelco

1-Methoxy-2-propanol

EMPLURA®

Synonym(s):

1-Methoxy-2-propanol, Propylene glycol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
90.12
MDL number:
UNSPSC Code:
12352103
EC Index Number:
203-539-1
NACRES:
NB.21
Assay:
≥99.5%
bp:
120 °C/1013 mbar
vapor pressure:
11.5 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

11.5 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.5%

form

liquid

autoignition temp.

287 °C

potency

>5000 mg/kg LD50, oral (Rat)
11000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-11.5 % (v/v)

impurities

≤0.002% Free acid (as C2H5COOH)
≤0.1% Water
≤0.29% 2-Methoxypropan-1-ol (GC)
≤100 ppb Fe (Iron)
≤1000 ppb Na (Sodium)
≤20 ppb Al (Aluminium)
≤20 ppb Cu (Copper)
≤20 ppb Ni (Nickel)
≤20 ppb Zn (Zinc)
≤50 ppb Ca (Calcium)
≤50 ppb K (Potassium)

pH

4-7 (20 °C, 200 g/L in H2O)

bp

120 °C/1013 mbar

mp

-97 °C

transition temp

flash point 31 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-25°C

SMILES string

O(CC(O)C)C

InChI

1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3

InChI key

ARXJGSRGQADJSQ-UHFFFAOYSA-N

General description

EMPLURA®
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application



  • Insights into the mechanism of the solvolysis of propylene oxide over titanium silicalite-1: a theoretical study.: This research offers a theoretical exploration of the solvolysis mechanisms of propylene oxide facilitated by titanium silicalite-1, contributing valuable insights into the catalytic processes involving similar volatile organic compounds (Qin et al., 2023).


  • Measuring the quantity of harmful volatile organic compounds inhaled through masks.: This study evaluates the levels of harmful volatile organic compounds, including 1-methoxy-2-propanol, that are inhaled through various types of masks, highlighting the potential health risks and the effectiveness of masks in blocking organic vapors (Ryu and Kim, 2023).



Analysis Note

Purity (GC)≥ 99.5 % (a/a)2-Methoxypropan-1-ol (GC)≤ 0.29 % (a/a)Identity (IR)passes testFree acid (as C₂H₅COOH)≤ 0.002 %Al (Aluminium)≤ 20 ppbCa (Calcium)≤ 50 ppbCu (Copper)≤ 20 ppbFe (Iron)≤ 100 ppbK (Potassium)≤ 50 ppbNa (Sodium)≤ 1000 ppbNi (Nickel)≤ 20 ppbZn (Zinc)≤ 20 ppbWater≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service