Skip to Content
Merck
All Photos(1)

Key Documents

1.15924

Sigma-Aldrich

Nigrosine (C.I. 50420)

water-soluble for microscopy Certistain®

Synonym(s):

Nigrosine (C.I. 50420)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116130
EC Index Number:
309-930-4

Quality Level

form

solid

pH

10 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

750 kg/m3

εmax

200-300 at 570-580 nm in 50% ethanol at 0.025 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Nigrosin (C.I. 50420) water-soluble for microscopy Certistain®, is used for the preparation of a staining solution for the visualization of capsule (negative capsule staining) in bacteriological material of human origin. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50 %): 570 - 580 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.025 g/l; ethanol 50 %): 200 - 300
TLC-Test: passes test
Loss on drying (110 °C): ≤ 15 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service