Skip to Content
Merck
All Photos(1)

Key Documents

1.14846

Supelco

Phosphate Test

colorimetric, 0.2-3.0 mg/L (PO4-P), for use with MQUANT®

Synonym(s):

Phosphate Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
€258.00

€258.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
€258.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€258.00


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

specific analyte(s)

phosphate

Quality Level

measuring range

0.2-3.0 mg/L (PO4-P)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, PMB, with color-disk comparator 0.2 - 0.4 - 0.6 - 0.8 - 1.0 - 1.5 - 2.0 - 2.5 - 3.0 mg/l PO4-P MQuant®
The MQuant® Phosphate test (formally MColortest®) can be used for the determination of orthophosphate in:
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Industrial and process water
  • Wastewater
  • Soils and fertilizers after appropriate sample pretreatment
  • Food after appropriate sample pretreatment
  • Steels after appropriate sample pretreatment

Method
In sulfuric solution orthophosphate ions react with molybdate ions to form molybdophosphoric acid. Ascorbic acid reduces this to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation. The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18465 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Application



  • Effects of Two Remineralizing Agents in Combination with Er:YAG and CO(2) Laser Irradiation on Microhardness of Demineralized Enamel: This study highlights the use of phosphate tests in dental research, specifically in measuring the efficacy of remineralizing agents combined with laser treatment to enhance enamel hardness, which is crucial for dental health and preventive care (Mapar M et al., 2024).


  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: This research utilizes phosphate tests in the study of plant physiology, specifically to gauge the efficacy of nano-fertilizers in enhancing phosphate uptake under salt stress conditions, providing insights into sustainable agricultural practices (Shoukat A et al., 2024).


  • Investigation of Mechanical Properties of Polymer-Infiltrated Tetrapodal Zinc Oxide in Different Variants: Applies phosphate testing in the field of materials science to assess the interaction of phosphate with novel composite materials, thereby informing the development of more durable and efficient materials for various industrial applications (Scherer F et al., 2024).


Analysis Note

Application testconforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service