1.10006
Nickel Test Strips, colorimetric
10-500 mg/L (Ni), MQuant®
Synonym(s):
Nickel chemical test strips
About This Item
Recommended Products
product name
Nickel Test, colorimetric, 10-500 mg/L (Ni), MQuant®
description
semi-quantitative
Quality Level
product line
MQuant®
specific analyte(s)
nickel
greener alternative product characteristics
Waste Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
measuring range
10-500 mg/L (Ni)
detection method
colorimetric
greener alternative category
storage temp.
15-25°C
General description
The MQuant® Nickel test is suitable for controlling waste water and galvanizing solutions, catalysts and for use in the glass and ceramics industry. It is also a very simple method of identifying metalic nickel in alloys or nickel plating. Nickel(II) ions react with dimethylglyoxime to form a red complex. The nickel concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the nickel concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® test.
We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.This product has benefited from:
- Test strip with reduction
- Recycled aluminum tube
- Digital instruction leaflet
Linkage
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service