Skip to Content
Merck
All Photos(1)

Key Documents

1.09023

Millipore

GRIESS-ILOSVAY′S nitrite reagent

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€85.30

€85.30


Estimated to ship on02 June 2025


Request more information

Select a Size

Change View
500 ML
€85.30

About This Item

UNSPSC Code:
41106212
NACRES:
NA.21

€85.30


Estimated to ship on02 June 2025


Request more information

form

liquid

Quality Level

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

pH

2.0 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

application(s)

environmental
food and beverages
food and beverages

microbiology

storage temp.

15-25°C

suitability

bacteria spp.

General description

Reagent solution for detecting microbial nitrite in order to identify nitrate-reducing microorganisms

Analysis Note

Appearance: clear, colourless to light pink
Sensitivity test: passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service