Skip to Content
Merck
All Photos(1)

Key Documents

1.08216

SAFC

Trehalose

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€51.00
100 G
€252.00

€51.00


Estimated to ship on30 May 2025


Request a Sample

Select a Size

Change View
10 G
€51.00
100 G
€252.00

About This Item

UNSPSC Code:
12352201

€51.00


Estimated to ship on30 May 2025


Request a Sample

Quality Level

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service