Skip to Content
Merck
All Photos(1)

Documents

1.08000

Sigma-Aldrich

Sputofluol

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

Quality Level

form

liquid

IVD

for in vitro diagnostic use

density

1.18 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Sputofluol - for microscopy, is used for human-medical cell diagnosis and serves the purpose of the microbiological investigation of sample material of human origin. It is a ready-to-use solution for the pretreatment of specimen material that when used together with other in vitro diagnostic products from our portfolio makes bacterial and mycotic target structures evaluable for diagnostic purposes (e. g. acid-fast bacteria (AFB)) by fixing, staining, counterstaining, mounting in microbiological specimen materials, for example smears of enriched bacterial cultures of e. g. the lung. The 100 ml bottle is sufficient for up to 1000 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

NaOH content (acidimetric): 7 - 8 %
Assay (iodometric, free Cl₂): ≥ 4.5 %
Specifications values and batch values refers to the time of produktion

Legal Information

SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service