Skip to Content
Merck
All Photos(2)

Key Documents

1.06051

Supelco

Dichloromethane

dried (max. 0.004% H₂O) SeccoSolv

Synonym(s):

DCM, Methylene chloride, Methylene dichloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-838-9

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

1223 °F
605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

contains

~50 ppm 2-methyl-2-butene as stabilizer

expl. lim.

22 %

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.004% Water
≤0.01% Carbon tetrachloride (GC)
≤0.01% Chloroform (GC)

evapn. residue

≤10 mg/L

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SeccoSolv ready-to-use dried solvents fulfill even the most stringent requirements. They are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency. To protect the quality of these products even better from potential contaminants, our new SeccoSept septum seal cap provides multiple layers of protection to keep solvents in flawless condition before, during, and after removal.

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Chloroform (GC): ≤ 0.01 %
Carbon tetrachloride (GC): ≤ 0.01 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.004 %
The product is stabilized with about 50 ppm 2-methyl-2-butene

Legal Information

SECCOSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service