Skip to Content
Merck
All Photos(1)

Key Documents

1.05562

Supelco

TLC plates, Silica gel 60 F₂₅₄

pkg of 1 roll, roll L × W 500 cm × 20 cm, aluminum support

Synonym(s):

F₂₅₄ Silica Gel TLC Plates

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€223.00

€223.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
€223.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

€223.00


Estimated to ship on01 June 2025


Request a Bulk Order

material

aluminum support
silica gel 60 matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 1 roll

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

200 μm

plate L × W

500 cm × 20 cm

roll L × W

500 cm × 20 cm

particle size

10-12 μm

pore size

60 Å medium pore diameter

storage temp.

2-30°C

General description

1 Aluminium roll 500 x 20 cm
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.

Application


  • Determination of β-blocking receptor drugs in silica gel TLC systems with the mobile phase containing surfactant.: This paper investigates the use of silica gel TLC plates with surfactant-containing mobile phases for the separation and quantitative analysis of β-blocking receptor drugs. The environmentally friendly method provides accurate LOD and LOQ values for various drugs (Traczuk and Polak, 2022).

  • Separation of peptides in thin-layer chromatography systems with C18 silica-based adsorbents.: This paper investigates the separation of peptides using thin-layer chromatography (TLC) systems with C18 silica-based adsorbents, including Silica gel 60 F254 plates. The study demonstrates the utility of these systems in peptide analysis (Gwarda et al., 2016).

Linkage

Replaces: 5562-7; 5562

Analysis Note

Specific surface area (according to BET; 5-Pt. measurement): 480 - 540 m²/g
Pore volume (N₂-isotherm): 0.74 - 0.84 ml/g
d 50 (laser diffraction, size distribution): 9.7 - 11.7 µm
Layer thickness: 160 - 210 µm
Deviation of layer thickness per plate: ≤ 40 µm
Chromatographic testing:
Colour test
hRf-values
- bleu vif organol, colour test, lipophile:
11 - 25
- Ceres Black G, colour test, lipophile: 34 - 48
- ceres Violet brn, colour test, lipophile: 52 - 67
Typical value determined on a conditioned sheet
Eluent: toluene (45% rel. humidity)

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service