Skip to Content
Merck
All Photos(1)

Documents

1.05110

Supelco

Potassium hexahydroxoantimonate(V)

cryst. for analysis EMSURE®

Synonym(s):

Potassium hexahydroxoantimonate(V), Potassium antimonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K[Sb(OH)6]
CAS Number:
Molecular Weight:
262.90
MDL number:
UNSPSC Code:
12352302
EC Index Number:
235-387-7
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.0% (calculated on calcined substance, iodometric)

form

solid

pH

7.5-9.0 (20 °C, 20 g/L in H2O)

solubility

20 g/L

bulk density

1200 kg/m3

cation traces

Na: ≤0.2%
excess alkali: ≤0.1% (as KOH)

storage temp.

2-30°C

InChI

1S/K.6H2O.Sb/h;6*1H2;/q+1;;;;;;;+3/p-6

InChI key

IDVSLFDKDQVCAP-UHFFFAOYSA-H

Related Categories

Analysis Note

Identity: passes test
Assay (iodometric, calculated on calcined substance): ≥ 99.0 %
Free alkali (as KOH): ≤ 0.1 %
Na (Sodium): ≤ 0.2 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service