1.05006
Bole white powder
powder, low bacterial content, for internal use
Synonym(s):
Bole white
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
form
powder
solid
pH
6-7 (20 °C, 50 g/L in H2O, slurry)
density
2.6 g/cm3 at 20 °C
application(s)
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
2-25°C
InChI
1S/2Al.O5Si2.2H2O.2O/c;;1-6(2)5-7(3)4;;;;/h;;;2*1H2;;/q2*+1;-2;;;;
InChI key
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
General description
Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Bole white (synonym kaolin) is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical applications, bole white has been used primarily for solid oral dosage forms as well as topical formulations.
Legal Information
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service