Skip to Content
Merck
All Photos(1)

Documents

1.04176

Sigma-Aldrich

Phloxin B (C.I. 45410)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid

Quality Level

potency

2870 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% gravimetric

pH

9.7 (20 °C, 1 g/L in H2O)

solubility

100 g/L

bulk density

650 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H4Br4Cl4O5.2Na/c21-5-1-3-17(9(23)15(5)29)32-18-4(2-6(22)16(30)10(18)24)20(3)8-7(19(31)33-20)11(25)13(27)14(28)12(8)26;;/h1-2,29-30H;;/q;2*+1/p-2

InChI key

OOYIOIOOWUGAHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Phloxin B (C.I. 45410) for microscopy Certistain®, is used for the preparation of a staining solution for cytoplasmic staining in histological sections of human origin. It is mainly used in Hemytoxylin and Eosin (H&E) overview staining that is routinely performed in histological laboratories. Eosin and Phloxine bind to the positively charged plasma proteins. The combination of both dyes results in a broader color range in the tissue. As a CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (gravimetrically): ≥ 80 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 546 - 550 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): 930 - 1400
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service