Skip to Content
Merck
All Photos(1)

Key Documents

1.03133

Supelco

Diantipyrylmethane

GR for analysis (reagent for titanium)

Synonym(s):

Diantipyrylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24N4O2
CAS Number:
Molecular Weight:
388.46
MDL number:
UNSPSC Code:
12352115
EC Index Number:
215-009-7

Quality Level

form

solid

mp

179-182 °C

solubility

0.431 g/L (experimental)

bulk density

430 kg/m3

storage temp.

15-25°C

InChI

1S/C23H24N4O2/c1-16-20(22(28)26(24(16)3)18-11-7-5-8-12-18)15-21-17(2)25(4)27(23(21)29)19-13-9-6-10-14-19/h5-14H,15H2,1-4H3

InChI key

QATHNKNPUVVKHK-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): conforms
Melting range: 179 - 182 °C
Loss on Drying (105 °C): ≤ 3.0 %
Suitability for titanium determination: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service