Skip to Content
Merck
All Photos(2)

Key Documents

1.02937

Supelco

N,N-Dimethylformamide

for spectroscopy Uvasol®

Synonym(s):

DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€107.00
2.5 L
€397.00

€107.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€107.00
2.5 L
€397.00

About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-679-5
NACRES:
NA.03
technique(s):
UV/Vis spectroscopy: suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

€107.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

833 °F

potency

3010 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

expl. lim.

15.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

270 nm, ≥25%
275 nm, ≥60%
290 nm, ≥80%
300 nm, ≥90%
330 nm, ≥98%

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

UV absorption

λ: 270 nm Amax: ≤0.60
λ: 275 nm Amax: ≤0.22
λ: 290 nm Amax: ≤0.10
λ: 300 nm Amax: ≤0.05
λ: 330 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Analysis Note

Purity (GC): ≥ 99.9 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.02 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 270 nm): ≥ 25 %
Transmission (at 275 nm): ≥ 60 %
Transmission (at 290 nm): ≥ 80 %
Transmission (at 300 nm): ≥ 90 %
Transmission (from 330 nm): ≥ 98 %
Absorbance (at 270 nm): ≤ 0.60
Absorbance (at 275 nm): ≤ 0.22
Absorbance (at 290 nm): ≤ 0.10
Absorbance (at 300 nm): ≤ 0.05
Absorbance (from 330 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according ACS

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service