Skip to Content
Merck
All Photos(1)

Key Documents

1.01534

Supelco

Silver sulfate

extra pure

Synonym(s):

Silver sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€331.00
250 G
€1,410.00

€331.00


Estimated to ship on22 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
50 G
€331.00
250 G
€1,410.00

About This Item

Empirical Formula (Hill Notation):
Ag2O4S
CAS Number:
Molecular Weight:
311.80
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-653-7
NACRES:
NB.21

€331.00


Estimated to ship on22 March 2025


Request a Bulk OrderRequest more information

Quality Level

potency

5000 mg/kg LD50, oral (Rat)

analyte functional class(es)

chemical oxygen demand (COD)

technique(s)

photometry: suitable

pH

5-6 (25 °C, 5 g/L in H2O)

mp

655 °C

solubility

8 g/L

density

5.45 g/cm3 at 20 °C

bulk density

1200 kg/m3

storage temp.

no temp limit

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.5 %
Identity: passes test
Chloride (Cl): ≤ 20 ppm
Cu (Copper): ≤ 50 ppm
Fe (Iron): ≤ 50 ppm
Pb (Lead): ≤ 50 ppm
Substances not precipitated by hydrochloric acid (as SO₄): ≤ 0.3 %

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service