Skip to Content
Merck
All Photos(1)

Documents

1.01350

Sigma-Aldrich

Leishman′s eosine methylene blue

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.21

Quality Level

form

solid

IVD

for in vitro diagnostic use

pH

5-6 (0.01% in methanol: water (1:1))

bulk density

700 kg/m3

εmax

650-850 at 522-525 nm in methanol at 0.0005%
800-1100 at 623-627 nm in methanol at 0.0005%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

General description

The dry mixture of the Leishman′s eosin methylene blue dye for microscopy, is used for the self-preparation of the Leishman′s staining solution. The staining method according to Leishman is one of the standard techniques in hematological diagnostic procedures. Like Wright′s eosin methylene blue and May-Grünwald′s eosin methylene blue, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The dye is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Identity (UV/VIS-Spectrum): passes test
pH-value (0,01 %; methanol 50 %): 5 - 6
Absorption maximum λ 1 (Methanol): 623 - 627 nm
Absorption maximum λ 2 (Methanol): 522 - 525 nm
Spec. Absorptivity A 1%/1cm
- λ1max; 0.0005 %; methanol; calculated on dried substance:
800 - 1100
- λ2max; 0.0005 %; methanol; calculated on dried substance: 650 - 850
TLC-Test: passes test
Loss on drying (105 °C): ≤ 8 %
Suitability for microscopy: passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service