Skip to Content
Merck
All Photos(1)

Key Documents

1.01189

Supelco

Lead borate

Lead borate. CAS 35498-15-8, EC Number 252-594-8, chemical formula PbxB(OH)₃.

Synonym(s):

Lead borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbxB(OH)3
CAS Number:
UNSPSC Code:
12352302
EC Index Number:
252-594-8

Quality Level

pH

8.6 (20 °C, 650 g/L in H2O)

density

11.0 g/cm3 at 20 °C

bulk density

3870 kg/m3

storage temp.

2-30°C

InChI

1S/2BO3.3Pb/c2*2-1(3)4;;;/q2*-3;3*+2

InChI key

XHBRTROQSUURDJ-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Residue on ignition: ≤ 0.5 %
Bulk density: 300 - 350 g/100 ml

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service