Skip to Content
Merck
All Photos(2)

Key Documents

1.01102

SAFC

Aluminium sulfate-18-hydrate

EMPROVE® ESSENTIAL Ph Eur,BP

Synonym(s):

Aluminium sulfate-18-hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 18H2O
CAS Number:
Molecular Weight:
666.43
UNSPSC Code:
12352302
EC Index Number:
233-135-0
form:
solid
solubility:
364 g/L

Agency

BP
Ph. Eur.

product line

EMPROVE® ESSENTIAL

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

pH

2.5-4.0 (20 °C, 20 g/L in H2O)

mp

90 °C

solubility

364 g/L

density

1.72 g/cm3

bulk density

820 kg/m3

InChI

1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6

InChI key

KFCWRPFNNIEZRY-UHFFFAOYSA-H

Analysis Note

Assay (complexometric , Al₂(SO₄)₃): 51.0 - 59.0 %
Identity: passes test
Appearance of solution: passes test
pH-value (2 %; water): 2.5 - 4.0
Chloride (Cl): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
As (Arsenic): ≤ 0.0003 %
Fe (Iron): ≤ 0.005 %
V (Vanadium)*: ≤ 0.0001
NH₄ (Ammonium): ≤ 0.01 %
Alkali- and alkaline earth metals: ≤ 0.4 %
Residual solvents (ICH Q3C): excluded by production process
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, BP

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service