Skip to Content
Merck

1.00664

SAFC

Parteck LUB CST

(Calcium stearate vegetable grade), EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF, FCC

Pharma Manufacturing

Synonym(s):

Parteck® LUB CST

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€112.00
20 KG
€1,054.00

€112.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
250 G
€112.00
20 KG
€1,054.00

About This Item

Linear Formula:
(CH3(CH2)16-COO)2Ca
CAS Number:
Molecular Weight:
607.02
MDL number:
UNSPSC Code:
12352107
EC Index Number:
216-472-8

€112.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest a Sample

Agency

BP
JP
NF
Ph. Eur.

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

fine powder

autoignition temp.

460 °C (DIN 51794)

potency

>10000 mg/kg LD50, oral (Rat)

particle size

~4 μm (d50)

mp

140-160 °C

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Ca+2].[O-]C(=O)CCCCCCCCCCCCCCCCC.[O-]C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Ca/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

CJZGTCYPCWQAJB-UHFFFAOYSA-L

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® LUB, a product line of our most effective excipients for lubrication, demonstrates excellent batch-to-batch consistency and helps you to achieve a reliable workflow in your tablet manufacture. Parteck® LUB CST is a vegetable-origin calcium stearate.

Features and Benefits

  • Consistent functionality and performance
  • Vegetable origin for minimal regulatory effort
  • Specified particle size and surface area for a reliable performance

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service