Skip to Content
Merck
All Photos(1)

Key Documents

1.00638

Millipore

Acrylamide-bis ready-to-use solution 40% (37.5:1)

for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
Pricing and availability is not currently available.

Quality Level

form

liquid

density

1.03 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay of N,N′-methylene-bisacrylamide (HPLC): 2.4 - 2.8 %
Appearance of solution (colour): almost colourless
Appearance of solution (clearness): almost clear
Free acid (as acrylic acid): ≤ 0.002 %
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.15

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service