Skip to Content
Merck
All Photos(3)

Key Documents

07-1225

Sigma-Aldrich

Anti-PP5 Antibody

Upstate®, from rabbit

Synonym(s):

Protein phosphatase T, Serine/Threonine phosphatase, Serine/threonine-protein phosphatase 5, Protein phosphatase 5, catalytic subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€296.00

€296.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€296.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€296.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP5C(5536)

General description

Protein phosphatase 5 (PP5) is a novel human protein serine/threonine phosphatase of molecular weight of 58 kDa. It is made up of a C-terminal phosphatase catalytic domain and an N-terminal domain, which has four repeats of 34 amino acids, three of which are tandemly arranged. The phosphatase domain contains all of the invariant motifs of the PP1/PP2A/PP2B gene family. However, it is not closely related to any other known member of this family of phosphatases. Thus it makes up a new subfamily of phosphatases. PP5 expressed in bacteria has been shown to dephosphorylate serine residues in proteins and is more sensitive than PP1 to the tumor promoter okadaic acid. PP5 appears to be localized to the nucleus where it is thought to play a roll in the regulation of RNA biogenesis and mitosis as part of a signaling pathway regulating cell cycle progression.

Specificity

Recognizes Protein Phosphatase 5 (PP5).

Immunogen

Synthetic peptide derived from the human protein phosphatase 5 protein.

Application

Detect PP5 using this Anti-PP5 Antibody validated for use in WB, IH(P).
Immunohistochemistry(paraffin):
Representative images from a previous lot.

Optimal Staining of PP5 Polyclonal Antibody: Human Testis
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Routinely evaluated by Western Blot on Jurkat lysates.

Western Blot Analysis: 1:500 dilution of this lot detected PP-5 on 10 μg of Jurkat lysates

Target description

58 kDa

Linkage

Replaces: AB4092

Physical form

Format: Purified
Purified rabbit polyclonal IgG in phosphate buffered saline with 0.08% sodium azide

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat Cell Lysate, A549 and MCF-7 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service