Skip to Content
Merck
All Photos(2)

Key Documents

05-731-I

Sigma-Aldrich

Anti-SH3KBPI Antibody, clone 179.1.E1

clone 179.1.E1, from mouse

Synonym(s):

SH3 domain-containing kinase-binding protein 1, Regulator of ubiquitous kinase, Ruk, SH3-containing expressed in tumorigenic astrocytes

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€115.00
100 μG
€310.00

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€115.00
100 μG
€310.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

179.1.E1, monoclonal

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

SH3 domain-containing kinase-binding protein 1 (UniProt: Q925Q9; also known as Regulator of ubiquitous kinase, Ruk, SH3-containing expressed in tumorigenic astrocytes, SH3KBPI,) is encoded by the Sh3kbp1 (also known as Ruk, Seta) gene (Gene ID: 84357) in rat. SH3KBPI is a peripheral membrane protein that serves as an adapter protein involved in regulating diverse signal transduction pathways. It localizes in endocytic vesicles containing clustered receptors. It is shown to be highly expressed in the spinal cord. It can self-associate and form homotetramers. It contains three SH3 domains (aa 1-58; 98-157; and 311-372). SH3KBPI is involved in the regulation of endocytosis and lysosomal degradation of ligand-induced receptor tyrosine kinases, including EGFR and MET/hepatocyte growth factor receptor, through an association with CBL and endophilins. It can attenuate phosphatidylinositol 3-kinase activity by interaction with its regulatory subunit. It is also reported to plays a role in the regulation of cell morphology and cytoskeletal organization. Seven different isoforms of SH3KBP1 have been described that are produced by alternative splicing.

Specificity

Clone 179.1.E1 is a mouse monoclonal antibody that specifically detects SH3 domain-containing kinase-binding protein 1.

Immunogen

His-tagged full-length recombinant rat SH3 domain-containing kinase-binding protein 1.

Application

Anti-SH3KBPI, clone 179.1.E1, Cat. No. 05-731-I, is a mouse monoclonal antibody that detects SH3 domain-containing kinase-binding protein 1 and has tested for use in Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot from this antibody detected SH3KBPI in NG108-15 cell lysate.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SH3KBPI in THP-1 cell lysate.

Target description

~85 kDa observed; 78.09 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service