Skip to Content
Merck
All Photos(1)

Key Documents

05-1579

Sigma-Aldrich

Anti-Axin-1 Antibody, clone A5

clone A5, from mouse

Synonym(s):

Axis inhibition protein 1, axin 1, axis inhibitor 1, fused, mouse, homolog of

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A5, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AXIN1(8312)
mouse ... Axin1(12005)

General description

Axin-1 is constitutively expressed and acts as a negative regulator of Wnt expression. It has been demonstrated that mouse Wnt, Wnt3a, is secreted by multiple cell lines and can act and as a growth factor for stem cells. Secreted Wnt3a is highly hydrophobic because it is palmitoylated on an essential cysteine C77; the palmitoylated state of Wnt3a is essential for stem cell signaling. Incubation of stem cells with Wnt3a in tissue culture medium resulted in the increased number of mouse hematopoietic stem cells expressing "primitive" phenotype.

Specificity

This antibody recognizes the N-terminus of Axin-1.

Immunogen

GST-tagged recombinant protein corresponding to the N-terminus of mouse Axin-1.

Application

Detect Axin-1 using this Anti-Axin-1 Antibody, clone A5 validated for use in WB, IP & IC.

Quality

Evaluated by Western Blot using immunoprecipitated Axin-1 in SW-480 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Axin-1 immunoprecipitated from 10 µg of SW-480 cell lysate.

Target description

~ 97 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuifeng Fan et al.
Molecular carcinogenesis, 54 Suppl 1, E138-E147 (2014-09-30)
Our previous studies indicate that abnormal expression of several Wnt signaling molecules including Axin, Dvl and β-catenin are involved in proliferation, invasion and metastasis of lung cancer. Zbed3 was found to inhibit function of Axin-GSK3β complex and thus lead to
Phosphatidylinositol 3-kinase signaling does not activate the wnt cascade.
Ng, SS; Mahmoudi, T; Danenberg, E; Bejaoui, I; de Lau, W; Korswagen, HC; Schutte, M; Clevers, H
The Journal of Biological Chemistry null
S Monteagudo et al.
Osteoarthritis and cartilage, 30(5), 724-734 (2022-03-02)
To investigate how ANP32A, previously linked to the antioxidant response, regulates Wnt signaling as unraveled by transcriptome analysis of Anp32a-deficient mouse articular cartilage, and its implications for osteoarthritis (OA) and diseases beyond the joint. Anp32a knockdown chondrogenic ATDC5 cells were
Rong Xie et al.
eLife, 11 (2022-12-22)
Axin1 is a key regulator of canonical Wnt signaling pathway. Roles of Axin1 in skeletal development and in disease occurrence have not been fully defined. Here, we report that Axin1 is essential for lower limb development. Specific deletion of Axin1
Eunsun Kim et al.
Nature communications, 10(1), 1909-1909 (2019-04-25)
Hepatocellular carcinoma (HCC) is the second leading cause of cancer-related deaths worldwide. β-catenin is widely thought to be a major oncogene in HCC based on the frequency of mutations associated with aberrant Wnt signaling in HCC patients. Challenging this model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service