Skip to Content
Merck
All Photos(1)

Key Documents

KEY466133674

Key Organics / BIONET

3-(3-methoxyphenoxy)butan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
194.23
MDL number:

Iupac Name

3-(3-methoxyphenoxy)-2-butanone

Inchi Code

1S/C11H14O3/c1-8(12)9(2)14-11-6-4-5-10(7-11)13-3/h4-7,9H,1-3H3

InChI key

VMULZYSJWXVFKN-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/12K-006

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/12K-006


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library