KEY309658693
Key Organics / BIONET
5,7-dichloro-3-ethylpyrazolo[1,5-a]pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5,7-dichloro-3-ethylpyrazolo[1,5-a]pyrimidine
Inchi Code
1S/C8H7Cl2N3/c1-2-5-4-11-13-7(10)3-6(9)12-8(5)13/h3-4H,2H2,1H3
InChI key
QFSOXHZZXZVMHY-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/SS-4332
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/SS-4332
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.