Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249CE21

J & W Pharmlab LLC

2-Iodo-1H-benzoimidazole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
280.5
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-iodo-1H-benzimidazole hydrochloride

Inchi Code

1S/C7H5IN2.ClH/c8-7-9-5-3-1-2-4-6(5)10-7;/h1-4H,(H,9,10);1H

InChI key

PQOINOLOPNYCQF-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/30R0018S

MSDS

https://www.jwpharmlab.com/home/product/30R0018S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library