Skip to Content
Merck
All Photos(1)

Key Documents

ENAH942F2F0E

Enamine

4-methyl-3,4-dihydro-2H-1,4-benzoxazin-8-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
164.21
MDL number:

Iupac Name

4-methyl-3,4-dihydro-2H-1,4-benzoxazin-8-ylamine

Inchi Code

1S/C9H12N2O/c1-11-5-6-12-9-7(10)3-2-4-8(9)11/h2-4H,5-6,10H2,1H3

InChI key

QNXQBHHHKHBBKT-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-4300982

MSDS

https://www.enaminestore.com/catalog/EN300-4300982


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library