Skip to Content
Merck
All Photos(1)

Documents

ENA018107182

Enamine

4-({2,2-dioxo-2lambda6-thia-3-azatricyclo[6.3.1.0,4,12]dodeca-1(11),4(12),5,7,9-pentaen-3-yl}methyl)benzohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
353.4
MDL number:

mp

155-156

Iupac Name

4-[(1,1-dioxido-2H-naphtho[1,8-cd]isothiazol-2-yl)methyl]benzohydrazide

Inchi Code

1S/C18H15N3O3S/c19-20-18(22)14-9-7-12(8-10-14)11-21-15-5-1-3-13-4-2-6-16(17(13)15)25(21,23)24/h1-10H,11,19H2,(H,20,22)

InChI key

ZAQRAQVNCROGOL-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-03677

Melting Point

155-156

MSDS

https://www.enaminestore.com/catalog/EN300-03677


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service