Skip to Content
Merck
All Photos(1)

Documents

ENA018105636

Enamine

2-chloro-1-[6-(2-chloroacetyl)-9H-fluoren-3-yl]ethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
319.19
MDL number:

mp

200-201

Iupac Name

1,1'-(9H-fluorene-3,6-diyl)bis(2-chloroethanone)

Inchi Code

1S/C17H12Cl2O2/c18-8-16(20)12-3-1-10-5-11-2-4-13(17(21)9-19)7-15(11)14(10)6-12/h1-4,6-7H,5,8-9H2

InChI key

RSLKYBMHESSAFB-UHFFFAOYSA-N

Purity

91

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-01880

Melting Point

200-201

MSDS

https://www.enaminestore.com/catalog/EN300-01880


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service