Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987EF030

ChemScene LLC (Preferred Partner)

2,3-Dihydrobenzofuran-7-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
148.16
MDL number:

bp

277.6+-29.0 C at 760 mmHg

mp

62-64C

Iupac Name

2,3-dihydro-1-benzofuran-7-carbaldehyde

Inchi Code

1S/C9H8O2/c10-6-8-3-1-2-7-4-5-11-9(7)8/h1-3,6H,4-5H2

InChI key

CLXXUGOCVBQNAI-UHFFFAOYSA-N

storage temp.

4C, stored under nitrogen

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/196799-45-8.html

Boiling point

277.6+-29.0 C at 760 mmHg

Melting Point

62-64C

Signal Word

Warning

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W005801.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library