C12H580D8855
Chemcia Scientific, LLC
6-Hydroxy-nicotinic acid tert-butyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate
Inchi Code
1S/C10H13NO3/c1-10(2,3)14-9(13)7-4-5-8(12)11-6-7/h4-6H,1-3H3,(H,11,12)
InChI key
ZQMWSXMQDAFZSO-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Light Yellow Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=5569
MSDS
http://www.chemcia.com/product_fl02.asp?id=5569
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.