C12H316CE98A
Chemcia Scientific, LLC
5-Bromo-3-iodo-imidazo[1,2-a]pyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-bromo-3-iodoimidazo[1,2-a]pyridine
Inchi Code
1S/C7H4BrIN2/c8-5-2-1-3-7-10-4-6(9)11(5)7/h1-4H
InChI key
HFOYQYSQYCGGBV-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Off-White Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=2445
MSDS
http://www.chemcia.com/product_fl02.asp?id=2445
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.