Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CE531

Chemcia Scientific, LLC

5,6,7,8-Tetrahydropyrido[3,4-c]pyridazin-3(2H)-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
187.63
MDL number:

Pricing and availability is not currently available.

Iupac Name

1,5,6,7-tetrahydropyrido[3,4-c]pyridazin-3(2H)-one hydrochloride

Inchi Code

1S/C7H9N3O.ClH/c11-7-3-5-1-2-8-4-6(5)9-10-7;/h3-4,8-9H,1-2H2,(H,10,11);1H

InChI key

ZMUZPUHKDDKEDJ-UHFFFAOYSA-N

storage temp.

0-5C

Salt Data

HCl

Purity

99%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Light Yellow Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=1172

MSDS

http://www.chemcia.com/product_fl02.asp?id=1172


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library