CH4424846074
ChemBridge Corporation
[4-(aminomethyl)benzyl]dimethylamine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-[4-(aminomethyl)benzyl]-N,N-dimethylamine dihydrochloride
Inchi Code
1S/C10H16N2.2ClH/c1-12(2)8-10-5-3-9(7-11)4-6-10;;/h3-6H,7-8,11H2,1-2H3;2*1H
InChI key
KYBFMFWHHZEYEP-UHFFFAOYSA-N
storage temp.
RT
Salt Data
2HCl
Purity
95%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
solid
Product Link
https://www.hit2lead.com/building-blocks/4014715
MSDS
https://www.hit2lead.com/sds/4014715_sds.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.