T-105
Testosterone Calibrator
Level 10, 2000 ( ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant®
Synonym(s):
Testosterone Calibrator
About This Item
packaging
pkg of 1 mL
manufacturer/tradename
Cerilliant®
IVD
for in vitro diagnostic use
concentration
2000 ( ng/dL, Stripped Serum)
format
single component solution
shipped in
dry ice
storage temp.
−20°C
General description
Legal Information
related product
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service